missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hydroxyapatite nanoparticles, 5-10% (w/v) aqueous colloidal dispersion, 20-50nm particles

Product Code. p-7070107
Click to view available options
Quantity:
25 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
This item is not returnable. View return policy

Product Code. 15485805

Brand: Thermo Scientific Chemicals J66691.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

20-50nm particles

Hydroxyapatite nanoparticles is used for polishing and dental powders, porcelain and pottery, and enameling. It is also utilized to engineer new biomaterials for applications such as bone grafts and fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Synonym nHAP; Nano- hydroxyapatite

Specifications

Content And Storage 2°C to 8°C
Format Liquid dispersion
Quantity 25 mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.