missing translation for 'onlineSavingsMsg'
Learn More
Hydroxylamine hydrochloride, ACS reagent, Thermo Scientific Chemicals Product Code.: 10361731

Hydroxylamine hydrochloride, ACS reagent, Thermo Scientific Chemicals

Product Code. 10361731
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10361731

Brand: Thermo Scientific Acros 412051000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hciShow MoreShow Less
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 155°C to 158°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 100 g
Linear Formula H2NOH·HCl
Fieser 01,478; 02,217; 07,176; 09,245
Merck Index 15, 4867
Solubility Information Solubility in water: 560g/L (20°C). Other solubilities: soluble in alcohol,methanol,glycerol,propylene glycol,insoluble in cold ether
Formula Weight 69.49
Percent Purity ≥96.0%
Grade ACS Reagent
Infrared Spectrum Authentic
Packaging Plastic bottle
Residue after Ignition 0.05% max
Chemical Name or Material Hydroxylamine hydrochloride, 96+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465455 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number412055000,412051000,412050025
Lot NumberA0463585 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number412055000,412051000,412050025
Lot NumberA0462610 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number412055000,412051000,412050025
Lot NumberA0458127 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number412055000,412051000,412050025
Lot NumberA0456631 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number412055000,412051000,412050025
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydroxylamine hydrochloride, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.