missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine-O-sulfonic acid, 97%
Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 210.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.97 |
| Packaging | Glass bottle |
| Linear Formula | H2NOSO3H |
| Quantity | 25 g |
| Fieser | 01,481; 02,217; 03,156; 04,256; 05,343; 06,290; 08,250; 09,245; 10,207; 15,17 |
| Solubility Information | Solubility in water: soluble |
| Formula Weight | 113.09 |
| Show More |
Safety and Handling
Product Identifier
- Hydroxylamine-O-sulfonic acid
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction