missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine-O-sulfonic acid, 97%

Product Code. 11404264
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11404264

Brand: Thermo Scientific Alfa Aesar A12560.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2950-43-8 | H3NO4S | 113.09 g/mol

Hydroxylamine-O-sulfonic acid is used in the synthesis of quinolones for the benzodiazepine site of GABAA receptors. It is also used as a polymerization catalyst and an amination reagent. It is involved in the preparation of diazene by reacting with an alkali. It is also used in the inhibition of polychlorinated dibenzo-p-dioxins (PCDD) as well as (polychlorinated biphenyl) PCB formation in co-combustion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxylamine-O-sulfonic acid is used in the synthesis of quinolones for the benzodiazepine site of GABAA receptors. It is also used as a polymerization catalyst and an amination reagent. It is involved in the preparation of diazene by reacting with an alkali. It is also used in the inhibition of polychlorinated dibenzo-p-dioxins (PCDD) as well as (polychlorinated biphenyl) PCB formation in co-combustion.

Solubility
Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in ether and chloroform.

Notes
Moisture sensitive and hygroscopic. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases, acid chlorides, acid anhydrides and corrodes metal.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2950-43-8
Molecular Formula H3NO4S
Molecular Weight (g/mol) 113.09
MDL Number MFCD00011604
InChI Key DQPBABKTKYNPMH-UHFFFAOYSA-N
Synonym hydroxylamine-o-sulfonic acid, aminooxy sulfonic acid, sulfoperamidic acid, hydroxylamine-o-sulphonic acid, permonosulfamic acid, amidoperoxymonosulfuric acid, aminooxysulfonic acid, amidosulfonic peracid, haos, sulfamic acid n-oxide
PubChem CID 76284
IUPAC Name amino hydrogen sulfate
SMILES NOS(O)(=O)=O

Specifications

Melting Point ∼210°C (decomposition)
Quantity 25 g
UN Number UN3260
Beilstein 956566
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol. Slightly soluble in ethanol. Insoluble in ether and chloroform.
Formula Weight 113.09
Percent Purity 0.97
Chemical Name or Material Hydroxylamine-O-sulfonic acid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine-O-sulfonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydroxylamine-O-sulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.