Learn More
Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals
£13.36 - £80.10
Chemical Identifiers
CAS | 10039-54-0 |
---|---|
Molecular Formula | H3NO·0.5H2SO4 |
MDL Number | MFCD00044869 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309359
|
Thermo Scientific Alfa Aesar
088944.14 |
25 g | |||||||
11379349
|
Thermo Scientific Alfa Aesar
088944.22 |
100 g | |||||||
11399349
|
Thermo Scientific Alfa Aesar
088944.36 |
500 g |
£32.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389349
|
Thermo Scientific Alfa Aesar
088944.A3 |
2 kg |
£80.10
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydroxylammonium sulfate is used to produce hydroxylamine-O-sulfonic acid from chlorosulfuric acid. It is widely used in organic synthesis for the conversion of aldehydes and ketones to oximes, carboxylic acids and their derivatives (e.g. esters) to hydroxamic acids, isocyanates to N-hydroxyureas and nitriles to amidoximes. It acts as a radical scavenger which is used to terminate the radical polymerization reactions. It finds application as an antioxidant in natural rubber. It is also used in photography as a stabilizer for color developers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10039-54-0 | |
H3NO·0.5H2SO4 | |
MFCD00044869 |
Specifications
10039-54-0 | |
99% | |
MFCD00044869 | |
Hydroxylamine sulfate |
Crystalline | |
H3NO·0.5H2SO4 | |
164.14 |
Safety and Handling
missing translation for 'einecsNumber' : 233-118-8
RUO – Research Use Only