missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine sulfate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 170.0°C |
Color | White |
Physical Form | Crystals |
Quantity | 5 kg |
Assay Percent Range | 99% |
Linear Formula | (H2NOH)2·H2SO4 |
Fieser | 01,481 |
Merck Index | 15, 4867 |
Solubility Information | Solubility in water: 329g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 164.14 |
Show More |
Safety and Handling
Product Identifier
- Hydroxylamine sulfate
- Warning
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H290-May be corrosive to metals.
- EUH044-Risk of explosion if heated under confinement.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'