missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylapatite, for analysis, Thermo Scientific Chemicals
£16.30 - £47.60
Chemical Identifiers
CAS | 1306-06-5 |
---|---|
Molecular Formula | Ca5HO13P3 |
Molecular Weight (g/mol) | 502.31 |
MDL Number | MFCD00010904 |
InChI Key | XYJRXVWERLGGKC-UHFFFAOYSA-D |
Synonym | Calcium phosphate hydroxide |
IUPAC Name | pentacalcium hydroxide triphosphate |
SMILES | [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10199372
|
Thermo Scientific Acros
371260250 |
25 g | Glass bottle |
£16.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10446583
|
Thermo Scientific Acros
371261000 |
100 g | Glass bottle |
£27.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10110273
|
Thermo Scientific Acros
371260010 |
1 kg | Plastic Bottle |
£47.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1306-06-5 | |
502.31 | |
XYJRXVWERLGGKC-UHFFFAOYSA-D | |
pentacalcium hydroxide triphosphate |
Ca5HO13P3 | |
MFCD00010904 | |
Calcium phosphate hydroxide | |
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O |
Specifications
1700.0°C | |
White | |
Ca5HO13P3 | |
MFCD00010904 | |
Calcium phosphate hydroxide | |
XYJRXVWERLGGKC-UHFFFAOYSA-D | |
pentacalcium hydroxide triphosphate | |
502.31 | |
Analytical | |
Hydroxylapatite |
1306-06-5 | |
Powder | |
3Ca3(PO4)2·Ca(OH)2 | |
15, 3514 | |
Solubility in water: negligible | |
[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O | |
502.31 | |
35 to 40% (Ca) | |
8% max. (800°C) |
Safety and Handling
missing translation for 'einecsNumber' : 215-145-7
RUO – Research Use Only