missing translation for 'onlineSavingsMsg'
Learn More

Hydroxynaphthol Blue, Indicator for Metal Titration, Honeywell Fluka™

Indicator for metal titration

£76.87 - £286.45

Chemical Identifiers

CAS 63451-35-4
Molecular Formula C20H11N2Na3O11S3
Molecular Weight (g/mol) 620.46
MDL Number MFCD00036393
InChI Key JQECMNGWLIFQLL-UHFFFAOYSA-K
PubChem CID 44134769
IUPAC Name trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15624110
View Documents Promotion Details
Honeywell-Fluka
33936-10G
10 g Glass Bottle
£76.87
10g
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
15634110
View Documents Promotion Details
Honeywell-Fluka
33936-50G
50 g Glass Bottle
£286.45
50g
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

63451-35-4
620.46
JQECMNGWLIFQLL-UHFFFAOYSA-K
trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
C20H11N2Na3O11S3
MFCD00036393
44134769
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

63451-35-4
MFCD00036393
JQECMNGWLIFQLL-UHFFFAOYSA-K
C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+]
620.46
620.47g/mol
Hydroxynaphthol blue
C20H11N2Na3O11S3
NONH for all modes of transport
Conforms to structure
trisodium;3-hydroxy-4-[2-(2-oxo-4-sulfonatonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonate
44134769
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : 264-197-7

SDS
Documents

Documents

Product Certifications