missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl cellulose, M.W. 100,000, Thermo Scientific Chemicals

Product Code. 11305687
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Thickener, binder, and emulsifier

Thickener, binder, and emulsifier.Hydroxypropyl cellulose is used as an emulsifier, stabilizer, whipping aid and thickener. It is also used as a binder in ceramics and glazes, additive in hair and cosmetic products and blow-molded bottles. Further, it is used in contact lens wetting solutions, as a formulation aid and texturizer for foods. In addition, it serves as a topical ophthalmic protectant and lubricant. It acts as a disintegrant and a binder in tablets. It is also utilized as a sieving matrix for DNA separations by capillary and microchip electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-64-2
Molecular Formula (C24H44O16)n
MDL Number MFCD00132688
Synonym 4-1-aminopropyl-n,n,3-trimethylaniline, 4-dimethylamino-alpha,2-dimethylphenethylamine, benzeneethanamine, 4-dimethylamino-alpha,2-dimethyl, amiflamin, 55875-51-9 di-hydrochloride, hydroxypropyl cellulose, m.w. 100,000, 4-1-aminopropyl-n,n,3-trimethyl-aniline, hydroxypropyl cellulose 6-10 mpa?s in water at 20 ?c, hydroxypropyl cellulose, 3-6 mpa?s in water at 20 ?c

Specifications

pH 7
Odor Odorless
Quantity 25 g
Merck Index 14,4841
Solubility Information Soluble in polar organic solvents,in water at room temperature;Soluble in water,methyl alcohol,ethyl alcohol and other organic solvents.
Physical Form Powder
Chemical Name or Material Hydroxypropyl cellulose, M.W. 100,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'