missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl β-Cyclodextrin, MP Biomedicals™

Product Code. 11428831
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Molecular biology and Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94035-02-6
Molecular Formula C54H102O39
Molecular Weight (g/mol) 1375.371
InChI Key BZRWCHBEBFPRGF-UHFFFAOYSA-N
Synonym 2-hydroxypropy-.beta.-cyclodextrin, 2-hydroxypropylether-b-cyclodextrin, hydroxypropyl beta-cyclodextrin, tetrakis 1 propanol ; @cyclo-hepta-amylose, cyclo-hepta-amylose; tetrakis propanol
PubChem CID 44134771
SMILES CCCO.CCCO.CCCO.CCCO.C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 1454
Physical Form Solid
Chemical Name or Material Hydroxypropyl β-Cyclodextrin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.