missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hydroxypropyl methyl cellulose, M.N. 86,000; viscosity 4,000mPas (2% solution)
£128.64 - £910.53
Chemical Identifiers
CAS | 9004-65-3 |
---|---|
Molecular Formula | C56H108O30 |
Molecular Weight (g/mol) | 1261.45 |
MDL Number | MFCD00131360 |
InChI Key | PUSNGFYSTWMJSK-GSZQVNRLSA-N |
PubChem CID | 57503849 |
IUPAC Name | (2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan- |
SMILES | CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10482811
|
Thermo Scientific Chemicals
244021000 |
100 g | Plastic bottle |
£128.64
100g |
In Stock
Log in to see stock. |
||||
10645652
|
Thermo Scientific Chemicals
244022500 |
250 g | Plastic bottle |
£276.38
250g |
In Stock
Log in to see stock. |
||||
10676404
|
Thermo Scientific Chemicals
244020010 |
1 kg | Plastic bottle |
£910.53
1kg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9004-65-3 | |
1261.45 | |
PUSNGFYSTWMJSK-GSZQVNRLSA-N | |
(2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan- |
C56H108O30 | |
MFCD00131360 | |
57503849 | |
CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
Specifications
White to Cream | |
Powder | |
MFCD00131360 | |
15, 4880 | |
PUSNGFYSTWMJSK-GSZQVNRLSA-N | |
(2R,3R,4S,5R,6R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane;1-[[(2R,3R,4S,5R,6S)-3,4,5-tris(2-hydroxypropoxy)-6-[(2R,3R,4S,5R,6R)-4,5,6-tris(2-hydroxypropoxy)-2-(2-hydroxypropoxymethyl)oxan- | |
57503849 | |
10% max. | |
Hydroxypropyl methyl cellulose |
9004-65-3 | |
C56H108O30 | |
1.3900g/mL | |
Solubility in water: soluble. Other solubilities: insoluble in hot water;, soluble in dmf, dmso, ethylene glycol (hot),, pyridine;, insoluble in anhydrous ethanol, diethyl ether,, chloroform | |
CC(COCC1C(C(C(C(O1)OC2C(OC(C(C2OCC(C)O)OCC(C)O)OCC(C)O)COCC(C)O)OCC(C)O)OCC(C)O)OCC(C)O)O.COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC | |
1261.45 | |
Authentic | |
1.39 |
Safety and Handling
missing translation for 'rtecsNumber' : NF9125000
missing translation for 'tsca' : TSCA
Product Identifier
- Hydroxypropyl methyl cellulose
RUO – Research Use Only