Learn More
Ansell HyFlex™ 11-135 Seamless Knit Polyamide Antistatic Gloves with Coated Fingertips
Gloves are knitted using a seamless, non-stop process with high-tenacity polyamid continuous thread. Antistatic properties protect users from electrostatic discharge.
£61.15
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Hand Specific
View More Specs
Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16059591
View Documents
Ansell
11135060
6
£61.15
12 pairs
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
16069591
View Documents
Ansell
11135070
7
£61.15
12 pairs
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
16079591
View Documents Promotion Details
Ansell
11135080
8
£61.15
12 pairs
In Stock
to see stock.
Add to basket
16089591
View Documents
Ansell
11135090
9
£61.15
12 pairs
In Stock
to see stock.
Add to basket
16099591
View Documents
Ansell
11135100
10
£61.15
12 pairs
Estimated Shipment: 24-02-2025
to see stock.
Add to basket
Description
Description
  • Gloves are knitted using a seamless, non-stop process
  • Textile liner made of high-tenacity polyamid continuous thread that includes copper
  • Seamless, fine stitches enhance user comfort
  • Stretchable polyurethane coating on the fingertips for grip capabilities
  • Antistatic properties offer the user hand protection against electrostatic discharge
  • Low lint for cleanroom applications
Specifications
Specifications
EN 420:2003 + A1:2009
Polyurethane
Specific
Anti-static Gloves
Unisex
11-135
Seamless Knit
Gray
Polyamide
113XA
Knit
13
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell HyFlex™ 11-135 Seamless Knit Polyamide Antistatic Gloves with Coated Fingertips