missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ HyFlex™ 11-300 (ex Monysoft 1)

Product Code. 15818747
Click to view available options
Size:
7
8
9
10
11
Product Type:
Light-duty Mechanical Protection Gloves
Mechanical Protection Gloves
This item is not returnable. View return policy
This item is not returnable. View return policy

Comfort and product protection

  • Glove knitted with a seamless non-stop process.
  • Textile liner made of polyamide thread with a high elasticity.
  • Elastic knitwrist.
  • Ambidextrous shape doubles the life of the glove by enabling the use of both sides.
  • Excellent adaptability owing to its extensibility.
  • Low lint glove.
  • Excellent comfort and dexterity
  • TRUSTED_SUSTAINABILITY

Specifications

Back Material Nylon
Classification Category I
Construction Seamless Knit
Cuff Material Nylon
Description Comfort and product protection
Color White
Disposable Reusable
Exterior Finish Nylon
Grade Industrial grade
Hand Specific
Latex Free Yes
Material Nylon
Powdered Powder Free
Product Type Mechanical Protection Gloves
Size 10
Sterility Non-sterile
Style Hyflex 11-300
For Use With (Application) Mechnical Protection
Glove Cuff Color White
Glove Cuff Style Knit
Hem Color Brown
Liner Material Nylon
Lining Knit Lining
Model 11-300
Packaging Type Polybag/cardboard box
Series Hyflex 11 series
Dimensions (L x W) 251 x 80 mm
Length (English) 251 mm
Weight (Metric) 9 g
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'