missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Hyflex™ 11-421 Series Light Duty Nitrile Polyurethane Gloves
Click to view available options
Size:
6
7
8
9
10
Description
- Multipurpose light duty gloves
- Palm fit coating
- Seamless black nylon liner, gauge 13
- Fine and stretchable stitches for excellent comfort
- Silicone-free
- Not suitable for contact with acidic foodstuffs
- Offers an indicator button (green) which belong to a range of color code corresponding to a cut level : 1 (green), 3 (orange), 5 (red)
Applications:
- Delicate manipulation
- Finishing works
- General handling
- Packaging
Specifications
Specifications
Certifications/Compliance | EN 420:2003 + A1:2009 and EN 388:2006 |
Description | Gauge: 13 |
Coating Material | Polyurethane |
Color | Gray, Black |
Exterior Finish | Palm Coated |
Material | Nitrile |
Product Type | Mechanical Protection Gloves |
Size | 8 |
Style | Construction: Knitted |
Liner Material | Nylon |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'