Learn More
Ansell™ HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves
Almost a second skin, this very thin glove offers outstanding comfort, fit and sensitivity for the most delicate handling. Ansell HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves feature 18-gauge knitting technology which allows the glove to be ultra-thin yet tough.
Brand:  Ansell™ 11619070
 View more versions of this product
Product Code. 12640732
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Designed for a wide range of lightweight applications in dry or slightly oily environments
  • Ideal for workers who need a higher level of fit, touch and precision than that offered by their current gloves
  • Also ideal for applications where cotton gloves or no gloves at all are used despite the risk of scrapes, blisters and minor injuries
  • Ideal for final assembly, airco, electrical engines, wire harness assembly, adjusters work
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyurethane
Polyurethane
Lightweight Gloves
11-619
Nylon
144 Pairs
CE Mark, EN 388 : 2121
Polyurethane
7
Knit
Coated-palm
11-619
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ansell™ HyFlex™ 11-619 Series Knitted Polyurethane Light Gloves > Size: 7