Learn More
Ansell Edmont™ HyFlex™ 11-630 Series Gray Polyurethane Medium Weight Gloves
Product Code. 11756663
Click to view available options
Size:
6
7
8
9
10
11
Yarn structure contains an optimal rate of HPPE. Offers high levels of cut resistance for a safe use in controlling, handling and assembly applications involving contacts with sharp pieces.
  • The Lycra™ in the liners of the gloves as well as the polyurethane coating provides excellent flexibility and fit.
  • The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
  • The PU formulation also increases abrasion resistance on the gloves’ work surface.
  • With strong cut resistance
  • Knitted construction
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 4443
Cuff Material Polyurethane
Product Type Medium Weight Gloves
Size 9
Glove Cuff Style Knitwrist
Liner Material Dupont™ Kevlar™ aramid fibers
Lining Nylon/Lycra™/Dyneema™/Cotton/Dupont™ Kevlar™ aramid fiber
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To High cut resistance