missing translation for 'onlineSavingsMsg'
Learn More
Ansell HyFlex™ 11-651 Medium Duty Cut-Resistant Industrial Gloves Product Code.: 16009641

Ansell HyFlex™ 11-651 Medium Duty Cut-Resistant Industrial Gloves

Product Code. 16009641
6
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy

Product Code. 16009641

Brand: Ansell 11651060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Intercept™ Cut Resistance Technology blends engineered and synthetic fibers into high performance yarns to provide cut protection, along with comfort and dexterity.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Constructed with a seamless, 13-gauge liner
  • Elastic knit wrist ensures comfort, dexterity and a secure fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Cuff Material Elastic
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Material Intercept Technology Yarn
Size 6
EN388 Standard 4X42C
For Use With (Application) Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-651
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell HyFlex™ 11-651 Medium Duty Cut-Resistant Industrial Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.