missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-751 Medium Duty Abrasion-Resistant Gloves, INTERCEPT™ Technology
GREENER_CHOICE

Product Code. p-7162786 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16059661

Brand: Ansell 11751060

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Level C cut protection gloves offer enhanced abrasion resistance for extended wear.

  • INTERCEPT Technology™ for high level of cut protection
  • High durability PU coating, increased levels of abrasion resistance
  • Dirt masking design allows for extended product use
  • In compliance with the standards EN420 (General requirements) and EN388 (Mechanical hazards)
  • Also available in vendor pack
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) CE, EN 420:2003 + A1:2009; Category II
Construction Knit
Coating Material Polyurethane
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HPPE, Fiberglass, Nylon, Spandex, Polyurethane
Product Type Cut-resistant Gloves
Size 6
EN388 Standard EN 388:4X43C
For Use With (Application) General maintenance, hardware assembly, machining, stamping, cutting, raw material handling
Glove Cuff Style Knit
Liner Material HPPE, Fiberglass, Nylon, Spandex
Model 11751060
Quantity 12 Pairs/Bag, 12 Bags/Cs.
Series HyFlex™ 11-751
Length (English) 8.8 in.
Length (Metric) 22.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.