missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell HyFlex™ 11-819 Nylon/Spandex ESD Gloves with FORTIX™ Nitrile Foam Coating
Safety gloves with ESD functionality certified to EN 16350:2014, protecting products against electrostatic discharge. Ultra-thin FORTIX™ coating enhances grip, while also maintaining bare hand sensitivity for tasks that require dexterity and precision.
£102.00
Specifications
Certifications/Compliance (Europe) | EN 16350:2014, EN 420:2003 + A1:2009 |
---|---|
Coated Area | Palm |
Applications | Industrial |
Coating Material | Nitrile Foam |
Color | Blue |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17270157
|
Ansell
11819060 |
6 |
£102.00
12 pairs |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
17280157
|
Ansell
11819070 |
7 |
£102.00
12 pairs |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
17290157
|
Ansell
11819080 |
8 |
£102.00
12 pairs |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
17200167
|
Ansell
11819090 |
9 |
£102.00
12 pairs |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
17210167
|
Ansell
11819100 |
10 |
£102.00
12 pairs |
In Stock
Log in to see stock. |
|||||
17220167
|
Ansell
11819110 |
11 |
£102.00
12 pairs |
Estimated Shipment: 03-04-2025
Log in to see stock. |
|||||
Description
- ESD functionality certified to EN 16350:2014
- Can be used in explosion hazard zones as part of a grounding chain
- Seamless, 18 gauge nylon and spandex liner helps maintain a bare-handed feel for tasks that require dexterity and precision
- FORTIX™ nitrile foam coating enhances grip and provides long lasting protection
- ZONZ™ Comfort Fit Technology ensures gloves are ultra breathable
- Proprietary washing process helps to deliver cleaner, more comfortable gloves
- Touchscreen functionality allows users to work with multiple touchscreen devices without glove removal
- Dermatologically approved by Dermatest™
- Certified by OEKO-TEX™
Specifications
EN 16350:2014, EN 420:2003 + A1:2009 | |
Industrial | |
Blue | |
Specific | |
Nylon, Spandex | |
Non-sterile | |
Aerospace, Automotive, Logistics, and Machinery/Equipment | |
Coated | |
18 |
Palm | |
Nitrile Foam | |
Reusable | |
Yes | |
ESD Gloves | |
3121A | |
Knit | |
HyFlex™ 11-819 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title