Learn More
Ansell™ HyFlex 11-840 Gloves Product Code.: 17059193
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Ansell™ HyFlex 11-840 Gloves
Product Code. 17059193
5
Click to view available options
Size:
5
12
Product Code. 17059193
Brand: Ansell™ 11840050
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
Add to basket
Mechanical protection gloves with nitrile foam coating on 15 gauge nylon-spandex liner, palm coated.
  • These industrial hand gloves offer Fortix™ Abrasion Resistance Technology for enhanced durability
  • Ideal nitrile-coated work gloves with up to 2x the grip for safer handling
  • Enhanced breathability for even better comfort
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Chlorinated No
Construction Knit
Coating Material Nitrile Foam
Color Black, Gray
Cut Resistance Rating ANSI A1
Exterior Finish Rough
Hand Ambidextrous
Latex Free Yes
Material Nylon and Spandex
Product Type Foam Nitrile Gloves
Size 5
Style 11-840
Glove Cuff Style Knit
Grip Design Ansell Grip Technology
Puncture Rating ANSI/ISEA 105-16 Level 3
Thumb Style Wing
Length (Metric) 200 to 275 mm
Product Title
Ansell™ HyFlex 11-840 Gloves >