missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Hyflex™ 11- 840 Series Light Duty Foam Nitrile Gloves
GREENER_CHOICE

Product Code. 15349069 Shop All Ansell Products
10
Click to view available options
Size:
10
11

Provides extreme durability and superior fit. Ansell HyFlex™ 11- 840 Series Light Duty Foam Nitrile Gloves are used for precision handling in abrasive applications.

  • Multipurpose light duty gloves
  • Tough 1.1mm FORTIX™ nitrile foam coating offers EN Abrasion Level 4 protection, with up to two times the abrasion performance of other lightweight, knit-dipped gloves
  • FORTIX™ coating is 20% more breathable than earlier formulations, for cooler, drier hands and less sweat
  • Matching the natural contours of the hand, the snug, second-skin liner is tailored to create more comfortable wearing experience: at the base of the little finger (added comfort), across the fingertips (superior tactility), and through the palm (greater dexterity)
  • Has balance of robust abrasion performance, enhanced comfort and high dexterity
  • Ideal solution for workers who move between tasks
  • Silicone-free material means no transfer of silicone contaminants to metal parts prior to painting

Applications:

  • Assembly of small and medium parts
  • Feeding the lines
  • General handling
  • Raw materials reception
  • Screwing and unscrewing
Greener Choice Claims
  • Environmental benefits include:
    • Free-of a substance that causes environmental harm
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:1999
Description Construction: Knitted
Color Black, Gray
Exterior Finish Palm Coated
Material Foam Nitrile
Product Type Light-duty Multipurpose Gloves
Size 10
Style Gauge: 15
Glove Cuff Style Knit
Liner Material Spandex Nylon
Series 11-840
Length (Metric) 200 to 270 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Hyflex™ 11- 840 Series Light Duty Foam Nitrile Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.