Learn More
Ansell™ HyFlex™ 11-920 Series Blue Nitrile Medium Weight Gloves
Product Code. 11706673 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 11706673
Brand: Ansell™ 11920080
Only null left
Add to basket
Add to basket
Handle small and medium-sized components lightly coated with oil or lubricant with confidence. Protects skin from oil contamination.
  • Patented Ansell Grip Technology™.
  • Microscopic channels in the glove coating guide away the oil for secure glove-to-surface contact.
  • Knitted construction
  • Nylon liner
  • Nitrile coating
  • Palm-coated finish
  • Knitwrist cuff
  • Blue color
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance ANSI, EN 388 : 3131
Cuff Material Nitrile
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Nylon
Product Type Mediumweight Gloves
Size 8
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Palm Type Coated-palm
Quantity 144 Pairs
Resistant To Good snag, puncture and superior abrasion resistance
Series 11-920