Learn More
Ansell™ HyFlex 11-944 Gloves
Click to view available options
Size: 7; Color: White; Material: Nylon
Description
- An ideal alternative to light leather gloves.
- Fine and stretchable stitches for excellent comfort.
- Designed specifically for use in moderately oily manufacturing and maintenance environments where protection combined with abrasion resistance are essential.
Specifications
Coating Material | Nitrile |
Color | White, Black |
Cut Resistance Rating | ANSI A1 |
Hand | Ambidextrous |
Material | Nylon |
Size | 7 |
Style | 11-944 |
Glove Cuff Style | Knit |
Length (Metric) | 260 mm |