missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 72-400 Lightweight Cut-Resistant Spandex Gloves

Product Code. 16825430
Click to view available options
Size:
6
7
8
9
10
11

Gloves provide a high level of resistance against cuts and abrasions without sacrificing dexterity. Lightweight design provides softness and flexibility so that gloves can be worn comfortably for an extended period of time.

  • Lightweight design provides softness and flexibility so that gloves can be comfortably worn for extended periods of time
  • Spandex elastic maintains a snug fit, even after laundering
  • Ambidextrous glove design extends useful life
  • Machine washable with non-caustic cleaners without affecting performance or durability
  • Compliant with FDA food handling requirements
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 407:2004, EN 511:2006, EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Material Intercept Technology Yarn
Size 11
EN388 Standard 3541F
For Use With (Application) Food Processing
Glove Cuff Style Knit
Series 72-400
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.