missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 72-400 Lightweight Cut-Resistant Spandex Gloves

Catalog Number 16875420
Click to view available options
Size:
6
7
8
9
10
11

Gloves provide a high level of resistance against cuts and abrasions without sacrificing dexterity. Lightweight design provides softness and flexibility so that gloves can be worn comfortably for an extended period of time.

  • Lightweight design provides softness and flexibility so that gloves can be comfortably worn for extended periods of time
  • Spandex elastic maintains a snug fit, even after laundering
  • Ambidextrous glove design extends useful life
  • Machine washable with non-caustic cleaners without affecting performance or durability
  • Compliant with FDA food handling requirements
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 407:2004, EN 511:2006, EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Material Intercept Technology Yarn
Size 6
EN388 Standard 3541F
For Use With (Application) Food Processing
Glove Cuff Style Knit
Series 72-400
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'