missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Hygiene Kit - QM24+
Click to view available options
For use with QM24+ Earmuffs
Description
- Extends performance and lifetime of earmuff device
- Recommended replacement every 6 months
- Snap-in ear cushions and foam earcup inserts
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Black |
For Use With (Equipment) | QM24+ Earmuffs |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'