missing translation for 'onlineSavingsMsg'
Learn More

HOPEN Hygiene PP Coverall with Collar

Product Code. 17266750
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Color:
Navy
White
This item is not returnable. View return policy
This item is not returnable. View return policy

Polypropylene hygiene coverall with zip fastener and collar

  • Hygiene PP Coverall
  • Classic with collar
  • Elasticated
  • 40 g/m2

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Material Polypropylene
Color White
Disposable Single-use
Sterility Non-sterile
Includes Collar
For Use With (Application) Controlled Environments
Style Elasticated
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'