Learn More
Sampling Systems™ Hygienic Butterfly Valve Kit
These hygienic butterfly valves are manufactured from 316L stainless steel and are ideal for many applications, including GMP use.
Brand:  Sampling Systems™ A136-01A
Additional Details : Weight : 1.70000kg
 View more versions of this product
Product Code. 17857817
  • £198.67 / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • Comes with a silicone or EPDM seal
  • Both seals are suitable for food and pharmaceutical use and come with relevant certification
  • Easy to operate: Simply turn the handle to open or close
  • Opening the handle slightly will mean you have reduced product flow
  • Valves are ideal for liquids
Specifications
Specifications
Valve Kit
Drum with Sloping Base
Non-sterile
Dispensing Liquid
Valve with EPDM Seals