missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hygromycin B, MedChemExpress
Click to view available options
Quantity:
200 mg
500 mg
1 g
5 g
Unit Size:
1g
200mg
500mg
5g
Description
Hygromycin B is an aminoglycoside antibiotic active against prokaryotic and eukaryotic cells.
Specifications
Specifications
| Chemical Name or Material | Hygromycin B |
| CAS | 31282-04-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H37N3O13 |
| Quantity | 200 mg |
| Synonym | Hygrovetine |
| Solubility Information | DMSO : 100 mg/mL (189.57 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 50 mg/mL (94.78 mM; Need ultrasonic) |
| SMILES | O[C@H]([C@H]1O)C2(O[C@@]([C@@H](N)CO)([H])[C@@H]1O)O[C@@]([C@@H](O[C@@H]3CO)O[C@@]([C@H]([C@H](N)C4)O)([H])[C@@H]([C@H]4NC)O)([H])[C@@]([C@H]3O)([H])O2 |
| Molecular Weight (g/mol) | 527.52 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction