missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hygromycin B, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Hygromycin B is an aminoglycoside antibiotic. Active against both prokaryotic and eukaryotic cells. Inhibits protein synthesis by binding to the 30S subunit and inhibiting the ribosomal translocation step of elongation.
Specifications
Specifications
Type | Hygromycin B |
CAS | 31282-04-9 |
Quantity | 50 mg |
Molecular Formula | C20H37N3O13 |
Synonym | O-6-Amino-6-deoxy-L-glycero-D-galacto-heptopyranosylidene-(1-2-3)-O-β-D-talopyranosyl(1-5)-2-deoxy-N3-methyl-D-streptamine |
InChI Key | GRRNUXAQVGOGFE-BBMONYMYSA-N |
SMILES | CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N |
IUPAC Name | (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol |
Molecular Weight (g/mol) | 527.524 |
PubChem CID | 134129613 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'