Learn More
HYI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559065
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALGCPRIHLM AGRVPQGADR IAVKAEMEAV FLENLRHAAG VLAQEDLVGL LEPINTRITD PQYFLDTPQQ AAAILQKVGR PNLQLQMDIF HWQIMDGNLT GNIREFLPIV GHVQVAQVPG RGEPSSPGEL NFPYLFQLLE DEGYKGFVGC Highest antigen sequence identity to the following orthologs: Mouse - 21%, Rat - 91%.
Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).Specifications
HYI | |
Polyclonal | |
Unconjugated | |
Hyi | |
2700033B16Rik; 6430559E15Rik; endothelial cell apoptosis protein E-CE1; HT036; hydroxypyruvate isomerase; hydroxypyruvate isomerase (putative); hydroxypyruvate isomerase homolog; hydroxypyruvate isomerase homolog (E. coli); HYI; Putative hydroxypyruvate isomerase; RGD1561416; SB156 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T013 | |
Hyi | |
Recombinant protein corresponding to Human HYI | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only