Learn More
Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Accessories
Product Code. p-6167031 Shop All Epredia Products
Click to view available options
Product Type:
Air Flow Indicator, Dwyer
Base Stand, Fixed
Drip Tray
Width (English):
36 in.
48 in.
N/A
For Use With (Equipment):
B1115361, B1115363, B1220361, B1220363
B1115361, B1115363, B1220361, B1220363, B1115362, B1115364, B1220362, B1220364
B1115482, B1115484, B1220482, B1220484
Unit Size:
Each
Product Code. 16661573
Brand: Epredia™ B1011482
This item requires a price update prior to being ordered.
Request a Price for this item here.
Add convenience and functionality to your fume hood with Epredia™ Hyperclean™ TruAir™ Ductless Fume Hood Accessories.
Accessory options include drip trays, base stands, air flow indicator, and filters.
Specifications
Product Type Drip Tray
Material Polypropylene
For Use With (Equipment) B1115482, B1115484, B1220482, B1220484
Width (English) 48 in.