Learn More
Corning™ HYPERFlask™ M Cell Culture Vessel
The High Yield PERformance Flask Manual (HYPERFlask™ M) vessel is specifically designed for manual use.
Brand:  Corning™ 10032
Product Code. 13489788
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Originally, the HYPERFlask™ cell culture vessel was developed for automation. Due to its popularity for use on the bench top, Corning™ has redesigned the HYPERFlask™ M vessel with new features.
  • Ergonomic Cap – smooth texture on liner cap allows more ergonomic closing and opening of vessel by operator
  • Adaptor Grid – new design allows for faster filling and emptying while reducing foam generation
  • Bar Code Serial Number – each individual flask is traceable by a serial number that can be read by the unaided eye or by a handheld bar code reader
  • Made with USP Class VI Materials – molded and assembled in a Certified Class 100,000 clean room
  • Sterile – gamma irradiated to sterility assurance level (SAL) 10−6
  • Clean Room Packaging – each pack is double bagged.
  • Innovative 10-Layer Design – 10 interconnected polystyrene growth surfaces
  • Growth Surface – Gas permeable polystyrene
  • Increase Cell Yield – 10 -fold higher cell yield increases productivity and capacity
  • Time and Space Savings – Reduce processing time and incubator storage space by handling one flask compared to 10 traditional 175 cm² flasks
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
4/Pk.
24
Sterile