missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum, 100L, Flat-bottom outer support container for BPCs

Use this LLDPE drum with 3-D bioprocess container systems and tank liners.

Brand:  Thermo Scientific™ SV50076.03

Code : 84

Additional Details : Weight : 1.00000kg

Product Code. 10195873

  • POA / Each
Estimated Shipment: 26-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

Use Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum with 3-D bioprocess container systems and tank liners.

  • Includes lid and clamps
  • Available in cylindrical, conical and flat-bottom
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications

Specifications

Flat-bottom Outer Support Container for Bioprocess Bags, Port detail 4 and 1.75 in.
Top Dispense
45 X 79 cm (17.63 x 31.25 in.)
100 L
HyPerforma™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers