Learn More
Thermo Scientific™ HyPerforma™ Mixtainer™ Hardware, Stainless-Steel Support Plate; EU
The Thermo Scientific™ Stain-less Steel Support Plate (EU) is part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications.
Brand: Thermo Scientific™ SV30097.03
Additional Details : Weight : 6.80388kg
Description
The Thermo Scientific™ Stain-less Steel Support Plate (EU) is part of the Thermo Scientific™ HyPerforma™ Mixtainer™ system to support bioproduction mixing applications. Additional components of the system include the Thermo Scientific™ HyPerforma™ Mixtainer™ BioProcess Container (BPC) with an integrated magnetic stir bar, a conical drum, a magnetic stirring plate and a dolly. The optimized support hardware allows mixing, moving, storage, shipping and discharge of the BPC contents while maintaining sterility of the contents. Standard products are available for sterile liquid-to-liquid or powder-to-liquid mixing.
Recommended for:
- Hydration of powdered media and buffers
- Sterile mixing of cell culture media with sera and reagents
- Sterile mixing of buffer solutions or other process fluids
- Pooling of sterile liquid fractions
Specifications
14.19 x 14.19 x 4 in. | |
Each | |
Stainless Steel (304) | |
HyPerforma™, Mixtainer™ |