Learn More
Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner
Product Code. p-7238142 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
19 L
28 L
38 L
378 L
757 L
Unit Size:
Pack of 10
Pack of 5
Product Code. 17810647
Brand: Thermo Scientific™ SH30647.10
Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
  • Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications:
  • Removes the need for tank cleaning and helps reduce cycle times.
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 378 L
Capacity (English) 100 gal.
Outer Packaging Cardboard Cartons
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9