missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperREZ™ XP Guard Cartridge Holder for HyperREZ XP 5.0 x 3.0mm guard cartridges

For use with Thermo Scientific™ HyperREZ XP 5.0 x 3.0mm Guard Cartridges.

Brand:  Thermo Scientific™ 60002-354

Product Code. 11517521

  • £348.74 / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Choose the Thermo Scientific™ HyperREZ™ XP Guard Cartridge Holder for use with HyperREZ XP 3.0 x 5.0mm Guard Cartridges.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 mm
5 mm
HyperREZ
Guard Cartridge
HyperREZ XP
Guard Cartridge Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.