Learn More
Thermo Scientific™ HyperSep™ C8 Plates
Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 well plates offer excellent performance.
£345.74 - £375.90
Specifications
For Use With (Application) | Screening of Drugs, Metabolites, and Peptides in Biological Samples |
---|---|
Particle Size | 40 to 60 μm |
Product Line | HyperSep |
Sorbent | HyperSep C8 |
Type | Solid Phase Extraction Well Plate |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13466986
|
Thermo Scientific™
60307-211 |
96-Well Plate; Bed weight 10 mg/2 mL |
£352.77
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
13496986
|
Thermo Scientific™
60307-212 |
96-Well Plate; Bed weight 25 mg/2 mL |
£368.87
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||||
13486986
|
Thermo Scientific™
60307-213 |
96-Well Plate; Bed weight 50 mg/2 mL |
£373.89
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
10513425
|
Thermo Scientific™
60300-445 |
96-Well Plate; Bed weight 10 mg |
£345.74
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
10283593
|
Thermo Scientific™
60300-446 |
96-Well Plate; Bed weight 25 mg |
£361.83
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
10485723
|
Thermo Scientific™
60300-447 |
96-Well Plate; Bed weight 50 mg |
£366.85
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
10015874
|
Thermo Scientific™
60300-448 |
96-Well Plate; Bed weight 100 mg |
£375.90
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep™ C8 Plates offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:
- Drugs and their metabolites in biological samples
- Peptides in biological samples
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
Specifications
Screening of Drugs, Metabolites, and Peptides in Biological Samples | |
HyperSep | |
Solid Phase Extraction Well Plate |
40 to 60 μm | |
HyperSep C8 | |
1 mL |