Learn More
Thermo Scientific™ HyperSep™ Cyano Cartridges
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE cartridges.
£171.86 - £325.62
Specifications
For Use With (Application) Retention of Polar Compounds From Nonpolar Matrices
For Use With (Equipment) Thermo Scientific HyperSep-96 System
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep Cyano
View More Specs
Products 8
Product Code Brand Bed Weight Column Capacity (Metric) Price Quantity & Availability  
Product Code Brand Bed Weight Column Capacity (Metric) Price Quantity & Availability  
10755564
View Documents
Thermo Scientific™
60108-745
100 mg 1 mL
£252.26
Pack of 100
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
10556375
View Documents
Thermo Scientific™
60108-746
50 mg 1 mL
£221.10
Pack of 100
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
10121083
View Documents
Thermo Scientific™
60108-747
200 mg 3 mL
£184.92
Pack of 50
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10765564
View Documents
Thermo Scientific™
60108-748
500 mg 3 mL
£171.86
Pack of 50
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
10364233
View Documents
Thermo Scientific™
60108-750
1000 mg 6 mL
£188.94
Pack of 30
Estimated Shipment: 20-06-2025
to see stock.
Add to basket
10239794
View Documents
Thermo Scientific™
60108-751
2000 mg 15 mL
£192.96
Pack of 20
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
10417134
View Documents
Thermo Scientific™
60108-752
5000 mg 25 mL
£325.62
Pack of 20
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
10630816
View Documents
Thermo Scientific™
60108-753
10 g 75 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep™ Cyano cartridges are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
  • Retention of polar compounds from hexane and oils
  • Reversed phase extraction of moderately polar compounds
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
Specifications
Retention of Polar Compounds From Nonpolar Matrices
40 to 60 μm
HyperSep Cyano
Thermo Scientific HyperSep-96 System
HyperSep
Solid Phase Extraction Cartridge
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Cyano Cartridges