Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
Brand:  Thermo Scientific™ 60105-304
 View more versions of this product
Product Code. 15230359
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Show More
Specifications
Specifications
150 mg MgSO4, 50 mg PSA, 50 mg C18
HyperSep
QuEChERS Clean-up
QuEChERS Method
Anhydrous Magnesium Sulfate, Primary/Secondary Amine, Endcapped C18
2 mL Centrifuge Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS) >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.