missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifolds

Product Code. p-4524468 Shop All Thermo Scientific Products
Click to view available options
Format:
16 port Vacuum Manifold
16-port vacuum manifold, for PFAS
24 port Vacuum Manifold
24-port vacuum manifold, for PFAS
For Use With (Application):
SPE
SPE, PFAS
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10447263

Brand: Thermo Scientific™ 60104232

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

16- and 24-port vacuum manifolds speeds up sample processing; glass block design allows visible inspection of the extraction process and simplifies cleaning.

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications. We offer a full range of extraction accessories to complement both traditional cartridges and 96-well plate formats.

Thermo Scientific™ HyperSep™ Glass Block Manifolds offer a simple way to quickly process multiple samples. The manifold's rigid Corian lid resists warping, and permits samples to elute directly into disposable test tubes. The design allows visible inspection of the extraction process and facilitates cleaning. HyperSep™ Glass Block Manifolds are compatible with the HyperSep™ Glass Block Vacuum Manifold Pump.

HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.

Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid

Vacuum control valve allows optimization of sample flow rates during extraction

Test tube racks allow the use of 12-13mm or 16mm disposable test tubes

Vacuum gauge and bleed valve enables easy monitoring of the vacuum process

Corner leg supports prevent damage to the luers and tips when the lid is not being used

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.

TRUSTED_SUSTAINABILITY

Specifications

Format 16 port Vacuum Manifold
For Use With (Application) SPE
For Use With (Equipment) Vacuum pump
Product Line HyperSep
Type Vacuum Manifold

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.