Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Adaptors Safety Tray
Use the safety tray as the bottom of the glass block vacuum manifold to contain spills.
Brand: Thermo Scientific™ 60104-260
This item is not returnable.
View return policy
Description
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Safety Tray as the bottom of the manifold and to contain spills. The HyperSep Glass Block Vacuum Manifold enables solid phase extraction sample preparation using 16 or 24 SPE cartridges.
Specifications
Manifold Safety Tray, 1 pk | |
Glass Block Vacuum Manifold Adaptors | |
Manifold Safety Tray |
SPE Sample Preparation | |
HyperSep |