Learn More
Thermo Scientific™ HyperSep™ Polypropylene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Brand: Thermo Scientific™ 60110-301
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polypropylene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
Specifications
C18; Polypropylene | |
40 to 60 μm | |
HyperSep C18 |
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
HyperSep | |
96-Well Plate |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.