Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
£286.00
Specifications
For Use With (Application) | Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules |
---|---|
Product Line | HyperSep |
Type | 96-Well Plate |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10289694
|
Thermo Scientific™
60110-201 |
C18; Polystyrene | N/A | ||||||
10723667
|
Thermo Scientific™
60110-206 |
Zirconium Dioxide; Polystyrene | N/A | ||||||
10138074
|
Thermo Scientific™
60110-207 |
Titanium Dioxide; Polystyrene | N/A | ||||||
10127364
|
Thermo Scientific™
60110-209 |
SAX; Polystyrene | N/A | ||||||
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
Specifications
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
HyperSep | |
96-Well Plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.