Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 1-10μL
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
£152.00 - £210.00
Specifications
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Product Line HyperSep
Type SPE Tips
Volume (Metric) 1 to 10 μL
View More Specs
Products 9
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10600184
View Documents
Thermo Scientific™
60109-201
BioBasic C18, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10703487
View Documents
Thermo Scientific™
60109-202
BioBasic C8, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10610184
View Documents
Thermo Scientific™
60109-203
BioBasic C4, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10456113
View Documents
Thermo Scientific™
60109-204
Hypercarb, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10098952
View Documents
Thermo Scientific™
60109-205
C18 plus Hypercarb, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10223233
View Documents
Thermo Scientific™
60109-206
HILIC, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10426754
View Documents
Thermo Scientific™
60109-207
Trypsin, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10059002
View Documents
Thermo Scientific™
60109-208
Titanium Dioxide, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10556005
View Documents
Thermo Scientific™
60109-217
Zirconium Dioxide, 1 to 10 μL Volume This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
  • Mass spectrometry
  • Desalting
  • Protein purification
  • MALDI
  • Electrophoresis
  • HPCE
  • HPLC
  • CEC
Tips are available in 1-10μL capacity and 10-200μL capacity.
Specifications
Specifications
Microscale Solid Phase Extraction
HyperSep
1 to 10 μL
Micropipettes
SPE Tips
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.