Learn More
Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE Cartridges.
Brand: Thermo Scientific™ 60303-408
Code : 16
Additional Details : Weight : 0.30000kg
Descrizione
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with quaternary amine functional groups. Their use in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Widely used in forensic and clinical analysis of THC and its metabolites, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.- High, consistent recoveries
- Ideal for large samples
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- pH stable 0 to 14
- Available in a wide range of chemistries and bed weights
Specifica
96-Well plate; Bed weight 60 mg | |
30 to 50 μm | |
HyperSep Retain AX |
Analysis of THC and its Metabolites | |
HyperSep | |
96-well Plate |