Learn More
Thermo Scientific™ HyperSep™ Retain PEP Cartridges
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
£99.41 - £461.00
Specifications
For Use With (Application) | Retention of Polar and Nonpolar Analytes |
---|---|
Product Line | HyperSep |
Sorbent | HyperSep Urea-Modified Polystyrene DVB |
Type | Solid Phase Extraction Cartridge |
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bed Weight | Column Capacity (Metric) | Price | Quantity & Availability | |||||
10595895
|
Thermo Scientific™
60107-201 |
30 mg | 1 mL |
£293.00
Pack of 100 |
Estimated Shipment: 17-04-2025
Log in to see stock. |
|||||
15974417
|
Thermo Scientific™
60107-225 |
600 mg | 6 mL |
£382.00
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10259564
|
Thermo Scientific™
60107-202 |
30 mg | 3 mL | N/A | ||||||
15984417
|
Thermo Scientific™
60107-226 |
300 mg | 3 mL | N/A | ||||||
10505905
|
Thermo Scientific™
60107-203 |
60 mg | 3 mL |
£134.16
Pack of 50 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
10448104
|
Thermo Scientific™
60107-204 |
200 mg | 3 mL |
£311.00
Pack of 50 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10351215
|
Thermo Scientific™
60107-205 |
500 mg | 3 mL |
£461.00
Pack of 50 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
|||||
10618725
|
Thermo Scientific™
60107-206 |
500 mg | 6 mL |
£272.36
Pack of 30 |
In Stock
Log in to see stock. ![]() |
|||||
10015894
|
Thermo Scientific™
60107-207 |
100 mg | 6 mL |
£199.00
Pack of 30 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
|||||
10269564
|
Thermo Scientific™
60107-208 |
60 mg | 6 mL |
£130.00
Pack of 30 |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
10279954
|
Thermo Scientific™
60107-211 |
150 mg | 6 mL |
£160.00
Pack of 30 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10565245
|
Thermo Scientific™
60107-212 |
200 mg | 6 mL |
£166.00
Pack of 30 |
In Stock
Log in to see stock. |
|||||
11504745
|
Thermo Scientific™
60107-214 |
2000 mg | 25 mL | N/A | ||||||
11514745
|
Thermo Scientific™
60107-215 |
1000 mg | 25 mL |
£359.00
Pack of 20 |
Estimated Shipment: 21-05-2025
Log in to see stock. |
|||||
13406936
|
Thermo Scientific™
60107-217 |
100 mg | 3 mL |
£171.00
Pack of 50 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
13436936
|
Thermo Scientific™
60107-218 |
1000 mg | 6 mL |
£426.00
Pack of 30 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features:Ideal for large samples, Hypersep Retain cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
- High, consistent recoveries
- Fast and easy sample preparation and method development
- Reproducible, reliable sample preparation in a traditional format
- Available in a wide range of chemistries and bed weights
- Drugs and metabolites in biological fluids
- Peptides in serum, plasma or biological fluids
- Environmental samples
Specifications
Retention of Polar and Nonpolar Analytes | |
HyperSep | |
HyperSep Urea-Modified Polystyrene DVB | |
Solid Phase Extraction Cartridge |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.