Learn More
Thermo Scientific™ HyperSep™ Solid Phase Extraction (SPE) Accessories, Base Plates
Base plates are used with Thermo Scientific™ HyperSep 96-well plates; other accessories include sample collection plates, adaptors and empty wells.
£110.13
Specifications
For Use With (Application) | SPE Sample Preparation |
---|---|
For Use With (Equipment) | HyperSep-96 Products and Compatible Manifolds |
Product Line | HyperSep |
Type | Base Plate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10649085
|
Thermo Scientific™
60300-303 |
5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep Base Plates are available singly or in packs of five, and are used with HyperSep 96-well plate applications.
HyperSep SPE Cartridges and PlatesHyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
Specifications
SPE Sample Preparation | |
HyperSep-96 Products and Compatible Manifolds | |
HyperSep | |
Base Plate |