Learn More
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
£280.00 - £455.00
Specifications
For Use With (Application) Microscale Solid Phase Extraction
For Use With (Equipment) Micropipettes
Product Line HyperSep
Type SpinTip
Volume (Metric) 10 to 200 μL
View More Specs
Products 10
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10404603
View Documents
Thermo Scientific™
60109-412
C18; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10535435
View Documents
Thermo Scientific™
60109-425
Zirconium Dioxide; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10140503
View Documents
Thermo Scientific™
60109-422
Titanium Dioxide; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10138004
View Documents
Thermo Scientific™
60109-421
Strong Cation Exchanger; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10498684
View Documents
Thermo Scientific™
60109-420
Strong Anion Exchanger; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10341145
View Documents
Thermo Scientific™
60109-419
Weak Anion Exchanger; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10426744
View Documents
Thermo Scientific™
60109-418
HyperSep SpinTip; 10-200 uL; Trypsin Material This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10314673
View Documents
Thermo Scientific™
60109-417
HILIC; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10716334
View Documents
Thermo Scientific™
60109-416
C18 plus Hypercarb; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10111273
View Documents
Thermo Scientific™
60109-415
Hypercarb; 10-200 uL Vol. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
Features:
Ideal for small sample volumes; features a 1 to 2μm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30μm).
  • Filter-free design to eliminate dead volume
  • No contamination from the supporting matrix
  • Available in many popular chemistries
Applications include:
  • Mass spectrometry
  • Desalting
  • Protein purification
  • MALDI
  • Electrophoresis
  • HPCE
  • HPLC
  • CEC
Specifications
Specifications
Microscale Solid Phase Extraction
HyperSep
10 to 200 μL
Micropipettes
SpinTip
Videos
SDS
Documents
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200μL