Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200μL
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Brand: Thermo Scientific™ 60109-216
Description
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
Specifications
Titanium Dioxide; 10 to 200 uL Volume | |
Micropipettes | |
HyperSep Titanium Dioxide | |
10 to 200 μL |
Microscale Solid Phase Extraction | |
HyperSep | |
SPE Tips |
Product Suggestions
Customers who viewed this item also viewed
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.